Carbamic acid, N-3-azetidinyl-, phenylmethyl ester 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Aminoazetidine, 3-CBZ protected
H52831.03
H52831.06
H52831.03EA
312
EUR
H52831.03
H52831.06
Carbamic acid, N-3-azetidinyl-, phenylmethyl ester 97%
Carbamic acid, N-3-azetidinyl-, phenylmethyl ester
Purity: 97%
CAS No.: 914348-04-2
Molecular Formula: C11H14N2O2
Formula Weight: 206.24
MDL No.: MFCD07368913
Formel:
C₁₁H₁₄N₂O₂ Molecular Weight: 206.25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368913 CAS-Nummer: 914348-04-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...