(2-(Methoxymethyl)phenyl)boronic acid 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (2-methoxymethylphenyl)boronic acid, 2-(Methoxymethyl)phenylboronic acid, 2-(Methoxymethyl)benzeneboronic acid, 2-Methoxymethylphenylboronic acid

H52858.MD H52858.03
H52858.MDEA 37.7 EUR
H52858.MD H52858.03
(2-(Methoxymethyl)phenyl)boronic acid 97%
(2-(Methoxymethyl)phenyl)boronic acid

Purity: 97% CAS No.: 126617-98-9 Molecular Formula: C8H11BO3 Formula Weight: 165.99 MDL No.: MFCD03412110

Formel: C₈H₁₁BO₃
Molecular Weight: 165,98 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03412110
CAS-Nummer: 126617-98-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR