5-Bromo-2-hydroxybenzeneboronic acid 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(5-Bromo-2-hydroxyphenyl)boronic acid, 5-Bromo-2-hydroxyphenylboronic acid
H52937.MD
H52937.03
H52937.MDEA
71.6
EUR
H52937.MD
H52937.03
5-Bromo-2-hydroxybenzeneboronic acid 96%
5-Bromo-2-hydroxybenzeneboronic acid
Purity: 97%
CAS No.: 89598-97-0
Molecular Formula: C6H6BBrO3
Formula Weight: 216.83
Physical Form: Solid
Appearance: White
Melting Point: 196-200°
MDL No.: MFCD00093408
Formel:
C₆H₆BBrO₃ Molecular Weight: 216.83 g/mol Schmelzpunkt: 196…200 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00093408 CAS-Nummer: 89598-97-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...