3-Carbamoyl-4-fluorobenzeneboronic acid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-(Aminocarbonyl)-4-fluorophenylboronic acid, (3-Carbamoyl-4-fluorophenyl)boronic acid
H53102.MD
H53102.03
H53102.MDEA
74.8
EUR
H53102.MD
H53102.03
3-Carbamoyl-4-fluorobenzeneboronic acid 97%
3-Carbamoyl-4-fluorobenzeneboronic acid
Purity: 97%
CAS No.: 874219-34-8
Molecular Formula: C7H7BFNO3
Formula Weight: 182.95
Melting Point: 257-259°
MDL No.: MFCD08235053
Formel:
C₇H₇BFNO₃ Molecular Weight: 182.95 g/mol Schmelzpunkt: 257…259 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08235053 CAS-Nummer: 874219-34-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...