(3-Fluoro-4-hydroxyphenyl)boronic acid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Fluoro-4-hydroxyphenylboronic acid, 3-Fluoro-4-hydroxybenzeneboronic acid
H53110.MD
H53110.03
H53110.MDEA
93
EUR
H53110.MD
H53110.03
(3-Fluoro-4-hydroxyphenyl)boronic acid 97%
(3-Fluoro-4-hydroxyphenyl)boronic acid
Purity: 97%
CAS No.: 182344-14-5
Molecular Formula: C6H6BFO3
Formula Weight: 155.92
Melting Point: 252-256°
MDL No.: MFCD06659838
Formel:
C₅H₅BFNO₂ Schmelzpunkt: 252…256 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659838 CAS-Nummer: 182344-14-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...