2-(Bromomethyl)benzeneboronic acid pinacol ester 98%

Lieferant: Thermo Fisher Scientific

Synonyme: 2-(2-(bromomethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-[2-(Bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(Bromomethyl)phenylboronic acid Pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl bromide

H53217.MD H53217.03
H53217.MDEA 36.8 EUR
H53217.MD H53217.03
2-(Bromomethyl)benzeneboronic acid pinacol ester 98%
2-(Bromomethyl)benzeneboronic acid pinacol ester

Purity: 98% CAS No.: 377780-72-8 Molecular Formula: C13H18BBrO2 Formula Weight: 297.00 Melting Point: 78-80° MDL No.: MFCD01632210

Formel: C₁₃H₁₈BBrO₂
Molecular Weight: 297.00 g/mol
Schmelzpunkt: 78…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01632210
CAS-Nummer: 377780-72-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR