4-N-(Boc)aminomethylphenylboronic acid 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: (4-(((tert-Butoxycarbonyl)amino)methyl)phenyl)boronic acid, 4-(tert-Butoxycarbonylaminomethyl)phenylboronic acid, 4-(Aminomethyl)benzeneboronic acid, N-BOC protected

H53225.MD H53225.03
H53225.MDEA 56.3 EUR
H53225.MD H53225.03
4-N-(Boc)aminomethylphenylboronic acid 95%
4-N-(Boc)aminomethylphenylboronic acid

Purity: 97% CAS No.: 489446-42-6 Molecular Formula: C12H18BNO4 Formula Weight: 251.09 Melting Point: 204-206° MDL No.: MFCD04115637

Formel: C₁₂H₁₈BNO₄
Molecular Weight: 251,09 g/mol
Schmelzpunkt: 204…206 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115637
CAS-Nummer: 489446-42-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR