E-Phenylethenylboronic acid 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
trans-2-Phenylvinylboronic acid
H53227.03
H53227.06
H53227.03EA
53.9
EUR
H53227.03
H53227.06
E-Phenylethenylboronic acid 97%
E-Phenylethenylboronic acid
Purity: 97%
CAS No.: 6783-05-7
Molecular Formula: C8H9BO2
Formula Weight: 147.97
Storage Temperature: -30°C to -10°C
Melting Point: 98-100°
MDL No.: MFCD00963621
Formel:
C₈H₉BO₂ Molecular Weight: 147,97 g/mol Schmelzpunkt: 98…100 °C |
MDL:
MFCD00963621 CAS-Nummer: 6783-05-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...