Aluminiumoxid superaktiviert, neutral Grade I

Lieferant: Thermo Scientific

45901.A1 45901.30
45901.A1EA 214 EUR
45901.A1 45901.30
Aluminiumoxid superaktiviert, neutral Grade I
Aluminium oxide superactivated neutral

CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 50-200 Micron APS Powder Refractive Index: 1.765 Appearance: White Odour: Odourless Melting Point: 2045° Boiling Point: 2980° Merck Reference: 14,356 MDL No.: MFCD00003424

Formel: Al₂O₃
Molecular Weight: 101,96 g/mol
Siedepunkt: 2980 °C
Schmelzpunkt: 2045 °C
Dichte: 3,94 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003424
CAS-Nummer: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 14,356

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR