Bis(3,5,6-trichloro-2-n-pentyloxycarbonylphenyl) oxalate 95%

Lieferant: Thermo Scientific

Synonyme: Bis(2-carbo-pentoxy-3,5,6-trichlorophenyl)oxalate

H53378.06 H53378.14
H53378.06EA 67.4 EUR
H53378.06 H53378.14
Bis(3,5,6-trichloro-2-n-pentyloxycarbonylphenyl) oxalate 95%
Bis(3,5,6-trichloro-2-n-pentyloxycarbonylphenyl) oxalate

Purity: 95% CAS No.: 75203-51-9 Molecular Formula: C26H24O8Cl6 Formula Weight: 677.19 Melting Point: 76-80° MDL No.: MFCD00012038

Formel: C₂₆H₂₄O₈Cl₆
Molecular Weight: 677,19 g/mol
Schmelzpunkt: 76…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012038
CAS-Nummer: 75203-51-9
EINECS: 278-124-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR