Ethyl-3-methyl-2-oxobutyrate 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Ethyl 3-methyl-2-oxobutanoate

H53413.06 H53413.14
H53413.06EA 59.6 EUR
H53413.06 H53413.14
Ethyl-3-methyl-2-oxobutyrate 97%
Ethyl-3-methyl-2-oxobutyrate

Purity: 97% CAS No.: 20201-24-5 Molecular Formula: C7H12O3 Formula Weight: 144.17 Flash Point: 43°(109°F) Refractive Index: 1.410 Boiling Point: 62°/11mm Beilstein Reference: 1756668 UN No.: UN3272 MDL No.: MFCD00009122

Formel: C₇H₁₂O₃
Molecular Weight: 144,17 g/mol
Siedepunkt: 62 °C (11 mmHg)
Dichte: 0,989
Flammpunkt: 43 °C (109 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009122
CAS-Nummer: 20201-24-5
EINECS: 243-587-0
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR