1-Methyl-N-carbobenzoxy-L-glutamate 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-Carbobenzoxy-L-glutamic Acid 1-Methyl Ester, Z-Glu-OMe, 1-Methyl N-Cbz-L-glutamate, (S)-4-(((Benzyloxy)carbonyl)amino)-5-methoxy-5-oxopentanoic acid, N-Cbz-L-glutamic Acid 1-Methyl Ester
H53417.06
H53417.03
H53417.06EA
355
EUR
H53417.06
H53417.03
1-Methyl-N-carbobenzoxy-L-glutamate 98%
1-Methyl-N-carbobenzoxy-L-glutamate
Purity: 98%
CAS No.: 5672-83-3
Molecular Formula: C14H17NO6
Formula Weight: 295.29
Refractive Index: 1.4365
Melting Point: 64-70°
Beilstein Reference: 2567599
Optical Rotation: -24° (c=1 in ethanol)
MDL No.: MFCD00083278
Formel:
C₁₄H₁₇NO₆ Molecular Weight: 295.29 g/mol Schmelzpunkt: 64…70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00083278 CAS-Nummer: 5672-83-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...