3-Iodo-1-phenylpropane 97%
Lieferant: Thermo Fisher Scientific
H53420.03
H53420.06
H53420.14
H53420.03EA
39.5
EUR
H53420.03
H53420.06
H53420.14
3-Iodo-1-phenylpropane 97%
3-Iodo-1-phenylpropane
Purity: 97%
CAS No.: 4119-41-9
Molecular Formula: C9H11I
Formula Weight: 246.09
Flash Point: 110°(230°F)
Refractive Index: 1.5820
MDL No.: MFCD00019036
Formel:
C₉H₁₁I Dichte: 1,530 Flammpunkt: 110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00019036 CAS-Nummer: 4119-41-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...