N-Benzylquininium chloride 95%

Lieferant: Thermo Fisher Scientific

Synonyme: N-Benzylquinine chloride, QUIBEC, (1S,2S,4S,5R)-1-Benzyl-2-((R)-hydroxy(6-methoxyquinolin-4-yl)methyl)-5-vinylquinuclidin-1-ium chloride

H53424.03 H53424.06
H53424.03EA 26.5 EUR
H53424.03 H53424.06
N-Benzylquininium chloride 95%
N-Benzylquininium chloride

Purity: 95% CAS No.: 67174-25-8 Molecular Formula: C27H31ClN2O2 Formula Weight: 451.01 Melting Point: 200-205° dec. Optical Rotation: -235° (c=1.5 in water) MDL No.: MFCD00198105

Formel: C₂₇H₃₁ClN₂O₂
Molecular Weight: 451.01 g/mol
Schmelzpunkt: 200…205 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00198105
CAS-Nummer: 67174-25-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR