5-Trimethylsilyl-4-pentyn-1-ol 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-(Trimethylsilyl)pent-4-yn-1-ol
H53457.06
H53457.14
H53457.06EA
116
EUR
H53457.06
H53457.14
5-Trimethylsilyl-4-pentyn-1-ol 97%
5-Trimethylsilyl-4-pentyn-1-ol
Purity: 97%
CAS No.: 13224-84-5
Molecular Formula: C8H16OSi
Formula Weight: 156.30
Flash Point: 84°(183°F)
Refractive Index: 1.457
Boiling Point: 76°/15mm
MDL No.: MFCD00190229
Formel:
C₈H₁₆OSi Molecular Weight: 156,30 g/mol Siedepunkt: 76 °C (15 mmHg) Dichte: 0,861 Flammpunkt: 84 °C (183 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00190229 CAS-Nummer: 13224-84-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...