1,1,3-Triphenyl-2-propin-1-ol 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1,1,3-Triphenylpropargyl alcohol, 1,1,3-Triphenylpropargyl Alcohol

H53461.06 H53461.14
H53461.06EA 49.1 EUR
H53461.06 H53461.14
1,1,3-Triphenyl-2-propin-1-ol 97%
1,1,3-Triphenyl-2-propyn-1-ol

Purity: 97% CAS No.: 1522-13-0 Molecular Formula: C21H16O Formula Weight: 284.36 Melting Point: 80-82° MDL No.: MFCD00004449

Formel: C₂₁H₁₆O
Molecular Weight: 284.36 g/mol
Schmelzpunkt: 80…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004449
CAS-Nummer: 1522-13-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR