3-Ethinylpyridin 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Pyridylacetylene
H53465.03
H53465.06
H53465.14
H53465.03EA
64.9
EUR
H53465.03
H53465.06
H53465.14
3-Ethinylpyridin 97%
3-Ethinylpyridin
Purity: 97%
CAS No.: 2510-23-8
Molecular Formula: C7H5N
Formula Weight: 103.12
Flash Point: 57°(135°F)
Melting Point: 39-40°
Boiling Point: 83-84°/30mm
UN No.: UN1325
MDL No.: MFCD02177459
Formel:
C₇H₅N Molecular Weight: 103,12 g/mol Siedepunkt: 83…84 °C (30 mmHg) Schmelzpunkt: 39…40 °C Flammpunkt: 57 °C (135 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD02177459 CAS-Nummer: 2510-23-8 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...