2-(Trifluoromethyl)phenylacetylene 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-Ethynyl-2-(trifluoromethyl)benzene

H53478.03 H53478.06
H53478.03EA 53.9 EUR
H53478.03 H53478.06
2-(Trifluoromethyl)phenylacetylene 97%
2-(Trifluoromethyl)phenylacetylene

Purity: 97% CAS No.: 704-41-6 Molecular Formula: C9H5F3 Formula Weight: 170.13 Flash Point: 25°(77°F) Refractive Index: 1.4760 Boiling Point: 45°/8mm UN No.: UN1993 MDL No.: MFCD00467313

Formel: C₉H₅F₃
Molecular Weight: 170,13 g/mol
Siedepunkt: 45 °C (8 mmHg)
Dichte: 1,222
Flammpunkt: 25 °C (77 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00467313
CAS-Nummer: 704-41-6
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR