Benzyl propargyl ether 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
((Prop-2-yn-1-yloxy)methyl)benzene
H53481.06
H53481.14
H53481.06EA
46.8
EUR
H53481.06
H53481.14
Benzyl propargyl ether 97%
Benzyl propargyl ether
Purity: 97%
CAS No.: 4039-82-1
Molecular Formula: C10H10O
Formula Weight: 146.19
Refractive Index: 1.5200
Boiling Point: 101-101°/18mm
MDL No.: MFCD00483987
Formel:
C₁₀H₁₀O Molecular Weight: 146,189 g/mol Siedepunkt: 101…101 °C (18 mmHg) Dichte: 1,011 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00483987 CAS-Nummer: 4039-82-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...