5,7-Dichloro-1-indanone 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
5,7-Dichloro-2,3-dihydro-1H-inden-1-one
H54007.03
H54007.06
H54007.03EA
103
EUR
H54007.03
H54007.06
5,7-Dichloro-1-indanone 97%
5,7-Dichloro-1-indanone
Purity: 97%
CAS No.: 448193-94-0
Molecular Formula: C9H6Cl2O
Formula Weight: 201.05
Melting Point: 98°
MDL No.: MFCD09908160
Formel:
C₉H₆Cl₂O Molecular Weight: 201.05 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09908160 CAS-Nummer: 448193-94-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...