2-(5-Norbornene-2,3-dicarboximido)-1,1,3,3-tetramethyluronium tetrafluoroborate ≥98%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: TNTU, 2-(1,3-Dioxo-3a,4,7,7a-tetrahydro-1H-4,7-methanoisoindol-2(3H)-yl)-1,1,3,3-tetramethyluronium tetrafluoroborate
H54194.06 H54194.14
H54194.06EA 122 EUR
H54194.06 H54194.14
2-(5-Norbornene-2,3-dicarboximido)-1,1,3,3-tetramethyluronium tetrafluoroborate ≥98%
2-(5-Norbornene-2,3-dicarboximido)-1,1,3,3-tetramethyluronium tetrafluoroborate
Purity: 98+% CAS No.: 125700-73-4 Molecular Formula: C14H20BF4N3O3 Formula Weight: 365.13 Storage Temperature: 2°C to 8°C Melting Point: 216-219° MDL No.: MFCD00167753
Formel: C₁₄H₂₀BF₄N₃O₃
Molecular Weight: 365.14 g/mol
Schmelzpunkt: 216…219 °C
MDL: MFCD00167753
CAS-Nummer: 125700-73-4