Isocytosin 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Amino-4(1H)-pyrimidinone, 2-Aminopyrimidin-4(1H)-one

H54228.03 H54228.06 H54228.14
H54228.03EA 37.7 EUR
H54228.03 H54228.06 H54228.14
Isocytosin 99%
Isocytosine

Purity: 99% CAS No.: 108-53-2 Molecular Formula: C4H5N3O Formula Weight: 111.10 MDL No.: MFCD00057557

Formel: C₄H₅N₃O
Molecular Weight: 111,10 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00057557
CAS-Nummer: 108-53-2
EINECS: 203-592-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR