N-Methoxy-N-methylbenzamide 98%

Lieferant: Thermo Fisher Scientific

H54253.06 H54253.14
H54253.06EA 74.4 EUR
H54253.06 H54253.14
N-Methoxy-N-methylbenzamide 98%
N-Methoxy-N-methylbenzamide

Purity: 98% CAS No.: 6919-61-5 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Flash Point: 113°(235°F) Refractive Index: 1.533 Boiling Point: 70° MDL No.: MFCD00075320

Formel: C₉H₁₁NO₂
Molecular Weight: 165.19 g/mol
Siedepunkt: 70 °C
Dichte: 1.085
Flammpunkt: 113 °C (235 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00075320
CAS-Nummer: 6919-61-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR