(1H-Indol-2-yl)methanamine 97%

Lieferant: Thermo Fisher Scientific
Achtung

H54281.MD H54281.03
H54281.MDEA 65.7 EUR
H54281.MD H54281.03
(1H-Indol-2-yl)methanamine 97%
(1H-Indol-2-yl)methanamine

Purity: 97% CAS No.: 21109-25-1 Molecular Formula: C9H10N2 Formula Weight: 146.19 MDL No.: MFCD03422512

Formel: C₉H₁₀N₂
Molecular Weight: 146.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03422512
CAS-Nummer: 21109-25-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR