Methyl-1H-pyrrole-3-carboxylate 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Methyl 1H-pyrrole-3-carboxylate

H54340.03 H54340.06
H54340.03EA 85.1 EUR
H54340.03 H54340.06
Methyl-1H-pyrrole-3-carboxylate 97%
Methyl-1H-pyrrole-3-carboxylate

Purity: 97% CAS No.: 2703-17-5 Molecular Formula: C6H7NO2 Formula Weight: 125.13 Melting Point: 85-89° MDL No.: MFCD00967074

Formel: C₆H₇NO₂
Schmelzpunkt: 85…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00967074
CAS-Nummer: 2703-17-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR