4-[4-(4'-Chloro-2-biphenylylmethyl)-1-piperazinyl]benzoic acid 97%
Lieferant: Thermo Scientific
|
Achtung
|
H54342.MD
H54342.03
H54342.MDEA
53.4
EUR
H54342.MD
H54342.03
4-[4-(4'-Chloro-2-biphenylylmethyl)-1-piperazinyl]benzoic acid 97%
4-[4-(4'-Chloro-2-biphenylylmethyl)-1-piperazinyl]benzoic acid
Purity: 97%
CAS No.: 916204-05-2
Molecular Formula: C24H23ClN2O2
Formula Weight: 406.91
MDL No.: MFCD16251290
Formel:
C₂₄H₂₃ClN₂O₂ Molecular Weight: 406,91 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16251290 CAS-Nummer: 916204-05-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...