2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one 97%
Lieferant: Thermo Scientific
Achtung
Synonyme: 2-Chloro-8-cyclopentyl-5-methylpyrido[2,3-d]pyrimidin-7(8H)-one
H54363.MD H54363.03 H54363.06
H54363.MDEA 51.2 EUR
H54363.MD H54363.03 H54363.06
2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one 97%
2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one
Purity: 97% CAS No.: 1013916-37-4 Molecular Formula: C13H14ClN3O Formula Weight: 263.72 Beilstein Reference: 15642863 MDL No.: MFCD13181207
Formel: C₁₃H₁₄ClN₃O
Molecular Weight: 263,72 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13181207
CAS-Nummer: 1013916-37-4