N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate ≥98%
Lieferant: Thermo Scientific
Synonyme: DABCYL acid, SE, DABCYL N-Succinimidyl Ester, 4-[4-(Dimethylamino)phenylazo]benzoic Acid N-Succinimidyl Ester, 2,5-Dioxopyrrolidin-1-yl 4-((4-(dimethylamino)phenyl)diazenyl)benzoate
H54402.MB H54402.MC
H54402.MBEA 61.1 EUR
H54402.MB H54402.MC
N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate ≥98%
N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate
Purity: 98+% CAS No.: 146998-31-4 Molecular Formula: C19H18N4O4 Formula Weight: 366.37 Storage Temperature: -30°C to -10°C MDL No.: MFCD00467415
Formel: C₁₉H₁₈N₄O₄
Molecular Weight: 366,38 g/mol
MDL: MFCD00467415
CAS-Nummer: 146998-31-4