4-(1,1'-Biphenyl-4-yl)-1,3-thiazol-2-amine 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-([1,1'-Biphenyl]-4-yl)thiazol-2-amine

H54413.MD H54413.03 H54413.06
H54413.MDEA 51.2 EUR
H54413.MD H54413.03 H54413.06
4-(1,1'-Biphenyl-4-yl)-1,3-thiazol-2-amine 97%
4-(1,1'-Biphenyl-4-yl)-1,3-thiazol-2-amine

Purity: 97% CAS No.: 2834-79-9 Molecular Formula: C15H12N2S Formula Weight: 252.33 Beilstein Reference: 187642 MDL No.: MFCD00047059

Formel: C₁₅H₁₂N₂S
Molecular Weight: 252,34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00047059
CAS-Nummer: 2834-79-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR