8-Methoxyquinoline 96%

Lieferant: Thermo Scientific

H54430.MD H54430.03 H54430.06
H54430.MDEA 42.9 EUR
H54430.MD H54430.03 H54430.06
8-Methoxyquinoline 96%
8-Methoxyquinoline

Purity: 96% CAS No.: 938-33-0 Molecular Formula: C10H9NO Formula Weight: 159.19 Melting Point: 38-41° MDL No.: MFCD00957068

Formel: C₁₀H₉NO
Molecular Weight: 159,19 g/mol
Schmelzpunkt: 38…41 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00957068
CAS-Nummer: 938-33-0
EINECS: 213-341-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR