4-Methyl-2-phenyl-1,3-thiazole-5-carboxylic acid 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Methyl-2-phenylthiazole-5-carboxylic acid

H54487.03 H54487.06
H54487.03EA 54.1 EUR
H54487.03 H54487.06
4-Methyl-2-phenyl-1,3-thiazole-5-carboxylic acid 97%
4-Methyl-2-phenyl-1,3-thiazole-5-carboxylic acid

Purity: 97% CAS No.: 33763-20-1 Molecular Formula: C11H9NO2S Formula Weight: 219.26 Melting Point: 202-206° MDL No.: MFCD00141955

Formel: C₁₁H₉NO₂S
Molecular Weight: 219.26 g/mol
Schmelzpunkt: 202…206 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00141955
CAS-Nummer: 33763-20-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR