N-(tert-Butoxycarbonyloxy)succinimide ≥98%
Lieferant: Thermo Scientific
Achtung
Synonyme: tert-Butyl (2,5-dioxopyrrolidin-1-yl) carbonate
H54493.06 H54493.14
H54493.06EA 120 EUR
H54493.06 H54493.14
N-(tert-Butoxycarbonyloxy)succinimide ≥98%
N-(tert-Butoxycarbonyloxy)succinimide
Purity: 98+% CAS No.: 13139-12-3 Molecular Formula: C9H13NO5 Formula Weight: 215.20 Storage Temperature: -30°C to -10°C Melting Point: ca 95° dec. MDL No.: MFCD00037903
Formel: C₉H₁₃NO₅
Molecular Weight: 215,20 g/mol
Schmelzpunkt: ∼ 95 °C (Zers.)
MDL: MFCD00037903
CAS-Nummer: 13139-12-3
EINECS: 236-071-1