Pentafluorophenyl trifluoroacetate ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Trifluoroacetic Acid Pentafluorophenyl Ester, Pentafluorphenyl trifluoracetate
H54529.14
H54529.06
H54529.14EA
294
EUR
H54529.14
H54529.06
Pentafluorophenyl trifluoroacetate ≥98%
Pentafluorophenyl trifluoroacetate
Purity: 98+%
CAS No.: 14533-84-7
Molecular Formula: C8F8O2
Formula Weight: 280.07
Flash Point: 52°(126°F)
Refractive Index: 1.368
Boiling Point: 122-123°
Beilstein Reference: 2003848
UN No.: UN3272
MDL No.: MFCD00134438
Formel:
C₈F₈O₂ Molecular Weight: 280.07 g/mol Siedepunkt: 122…123 °C Dichte: 1.63 Flammpunkt: 52 °C (126 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134438 CAS-Nummer: 14533-84-7 ADR: 3 III |