2-Pyrimidincarbonsäure 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Pyrimidine-2-carboxylic acid, 2-Carboxypyrimidine
H54745.03
H54745.06
H54745.03EA
96.9
EUR
H54745.03
H54745.06
2-Pyrimidincarbonsäure 97%
2-Pyrimidinecarboxylic acid
Purity: 97%
CAS No.: 31519-62-7
Molecular Formula: C5H4N2O2
Formula Weight: 124.10
MDL No.: MFCD00856161
Formel:
C₅H₄N₂O₂ Molecular Weight: 124.1 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00856161 CAS-Nummer: 31519-62-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...