2-Amino-3-methylpyridine-5-boronic acid pinacol ester 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H54773.MD
H54773.03
H54773.MDEA
151
EUR
H54773.MD
H54773.03
2-Amino-3-methylpyridine-5-boronic acid pinacol ester 96%
2-Amino-3-methylpyridine-5-boronic acid pinacol ester
Purity: 96%
CAS No.: 1111637-91-2
Molecular Formula: C12H19BN2O2
Formula Weight: 234.10
Storage Temperature: 2°C to 8°C
Melting Point: 128-130°
MDL No.: MFCD12923388
Formel:
C₁₂H₁₉BN₂O₂ Schmelzpunkt: 128…130 °C |
MDL:
MFCD12923388 CAS-Nummer: 1111637-91-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...