Fluor-N,N,N′,N′-tetramethylformamidiniumhexafluorphosphat 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
TFFH, N-((Dimethylamino)fluoromethylene)-N-methylmethanaminium hexafluorophosphate(V)
H54910.03
H54910.06
H54910.03EA
73.3
EUR
H54910.03
H54910.06
Fluor-N,N,N′,N′-tetramethylformamidiniumhexafluorphosphat 98%
Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate
Purity: 98%
CAS No.: 164298-23-1
Molecular Formula: C5H12F7N2P
Formula Weight: 264.12
Melting Point: 104-111°
MDL No.: MFCD02684443
Formel:
C₅H₁₂F₇N₂P Molecular Weight: 264,13 g/mol Schmelzpunkt: 104…111 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02684443 CAS-Nummer: 164298-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...