Ethyl-2-brom-1,3-thiazol-5-carboxylat 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ethyl 2-bromothiazole-5-carboxylate

H55109.03 H55109.06
H55109.03EA 86.6 EUR
H55109.03 H55109.06
Ethyl-2-brom-1,3-thiazol-5-carboxylat 98%
Ethyl-2-bromothiazole-5-carboxylate

Purity: 98% CAS No.: 41731-83-3 Molecular Formula: C6H6BrNO2S Formula Weight: 236.08 Flash Point: 110°(230°F) Refractive Index: 1.5530 Boiling Point: 216-217° MDL No.: MFCD00463837

Formel: C₆H₆BrNO₂S
Molecular Weight: 236.08 g/mol
Siedepunkt: 216…217 °C
Dichte: 1.581
Flammpunkt: 110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00463837
CAS-Nummer: 41731-83-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR