(S)-4-Cyano-4'-(2-methylbutyl)biphenyl 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (S)-4'-(2-Methylbutyl)-[1,1'-biphenyl]-4-carbonitrile

H55183.03 H55183.06
H55183.03EA 119 EUR
H55183.03 H55183.06
(S)-4-Cyano-4'-(2-methylbutyl)biphenyl 97%
(S)-4-Cyano-4'-(2-methylbutyl)biphenyl

Purity: 97% CAS No.: 63799-11-1 Molecular Formula: C18H19N Formula Weight: 249.36 MDL No.: MFCD00192280

Formel: C₁₈H₁₉N
Molecular Weight: 249,36 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00192280
CAS-Nummer: 63799-11-1
EINECS: 264-464-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR