Potassium tetraphenylborate 97%

Lieferant: Thermo Fisher Scientific
Achtung

H55276.06 H55276.14
H55276.06EA 67.1 EUR
H55276.06 H55276.14
Potassium tetraphenylborate 97%
Potassium tetraphenylborate

Purity: 97% CAS No.: 3244-41-5 Molecular Formula: C24H20BK Formula Weight: 358.33 Physical Form: Powder Appearance: White Melting Point: 300° MDL No.: MFCD03093877

Formel: C₂₄H₂₀BK
Molecular Weight: 358.33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03093877
CAS-Nummer: 3244-41-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR