Boc-Cha-OH 98%
Lieferant: Thermo Scientific
Synonyme:
(S)-2-((tert-Butoxycarbonyl)amino)-3-cyclohexylpropanoic acid, Boc-β-cyclohexyl-Ala-OH
H55282.MD
H55282.03
H55282.06
H55282.MDEA
38.4
EUR
H55282.MD
H55282.03
H55282.06
Boc-Cha-OH 98%
Boc-Cha-OH
Purity: 98%
CAS No.: 37736-82-6
Molecular Formula: C14H25NO4
Formula Weight: 271.36
Beilstein Reference: 3060603
MDL No.: MFCD00211335
Formel:
C₁₄H₂₅NO₄ Molecular Weight: 271,357 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00076896 CAS-Nummer: 37736-82-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...