4-Boc-1-(5-bromo-2-pyridyl)piperazine 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
5-Bromo-2-(4-tert-butoxycarbonylpiperazin-1-yl)pyridine, tert-Butyl 4-(5-bromopyridin-2-yl)piperazine-1-carboxylate
H55311.03
H55311.06
H55311.03EA
46.7
EUR
H55311.03
H55311.06
4-Boc-1-(5-bromo-2-pyridyl)piperazine 97%
4-Boc-1-(5-bromo-2-pyridyl)piperazine
Purity: 97%
CAS No.: 153747-97-8
Molecular Formula: C14H20BrN3O2
Formula Weight: 342.23
Physical Form: Powder
Appearance: White to yellow
Melting Point: 83-87°
UN No.: UN2811
MDL No.: MFCD07369772
Formel:
C₁₄H₂₀BrN₃O₂ Molecular Weight: 342.237 g/mol Schmelzpunkt: 83…87 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07369772 CAS-Nummer: 153747-97-8 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...