Di-tert-butyl-L-(+)-tartrate 99%
Lieferant: Thermo Scientific
Synonyme:
(2R,3R)-Di-tert-butyl 2,3-dihydroxysuccinate, L-(+)-Tartaric Acid Di-tert-Butyl Ester
H55860.MD
H55860.03
H55860.MDEA
185
EUR
H55860.MD
H55860.03
Di-tert-butyl-L-(+)-tartrate 99%
Di-tert-butyl-L-(+)-tartrate
Purity: 99%
CAS No.: 117384-45-9
Molecular Formula: C12H22O6
Formula Weight: 262.30
Flash Point: 109°(228°F)
Physical Form: Solid
Density: 1.143
Appearance: White
Melting Point: 90-92°
Beilstein Reference: 5026723
Optical Rotation: +11° (c=1 in acetone)
MDL No.: MFCD00192000
Formel:
C₁₂H₂₂O₆ Molecular Weight: 262,30 g/mol Schmelzpunkt: 90…92 °C Dichte: 1,143 Flammpunkt: 109 °C (228 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00192000 CAS-Nummer: 117384-45-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...