Phenyl(1-piperazinyl)methanon 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Phenyl(piperazin-1-yl)methanone

H55896.03 H55896.06
H55896.03EA 38.5 EUR
H55896.03 H55896.06
Phenyl(1-piperazinyl)methanon 97%
1-Benzoylpiperazine

Purity: 97% CAS No.: 13754-38-6 Molecular Formula: C11H14N2O Formula Weight: 190.24 Melting Point: 61-64° Boiling Point: 145°/0.05mm MDL No.: MFCD00810192

Formel: C₁₁H₁₄N₂O
Molecular Weight: 190,24 g/mol
Siedepunkt: 145 °C (0,05 mmHg)
Schmelzpunkt: 61…64 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00810192
CAS-Nummer: 13754-38-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR