3,4-Dihydro-2H-pyran-2-ylmethanol 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Hydroxymethyl-3,4-dihydro-2H-pyran, DHP Linker

H56124.03 H56124.06
H56124.03EA 38.5 EUR
H56124.03 H56124.06
3,4-Dihydro-2H-pyran-2-ylmethanol 96%
3,4-Dihydro-2H-pyran-2-ylmethanol

Purity: 96% CAS No.: 3749-36-8 Molecular Formula: C6H10O2 Formula Weight: 114.14 Storage Temperature: 2°C to 8°C Flash Point: 89°(192°F) Refractive Index: 1.4780 Boiling Point: 92-93°/25mm MDL No.: MFCD00051507

Formel: C₆H₁₀O₂
Molecular Weight: 114,14 g/mol
Siedepunkt: 92…93 °C (25 mmHg)
Dichte: 1,101
Flammpunkt: 89 °C (192 °F)
MDL: MFCD00051507
CAS-Nummer: 3749-36-8
EINECS: 223-150-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR