3-Aminooxetane-3-carboxylic acid 95%

Lieferant: Thermo Scientific
Achtung

H57291.MD H57291.ME
H57291.MDEA 139 EUR
H57291.MD H57291.ME
3-Aminooxetane-3-carboxylic acid 95%
3-Aminooxetane-3-carboxylic acid

Purity: 95% CAS No.: 138650-24-5 Molecular Formula: C4H7NO3 Formula Weight: 117.1 Physical Form: Powder Appearance: White Melting Point: 205-210° MDL No.: MFCD11035888

Formel: C₄H₇NO₃
Molecular Weight: 117,1 g/mol
Schmelzpunkt: 205…210 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11035888
CAS-Nummer: 138650-24-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR