Oxetane-3-carboxylic acid 95%

Lieferant: Thermo Fisher Scientific
Achtung

H57578.MDEA 410 EUR
H57578.MD H57578.ME
Oxetane-3-carboxylic acid 95%
Oxetane-3-carboxylic acid

Purity: 95% CAS No.: 114012-41-8 Molecular Formula: C4H6O3 Formula Weight: 102.09 Storage Temperature: -30°C to -10°C Flash Point: 115°(239°F) Boiling Point: 244° MDL No.: MFCD08669505

Formel: C₄H₆O₃
Molecular Weight: 102.09 g/mol
Siedepunkt: 244 °C
Dichte: 1.374
Flammpunkt: 115 °C (239 °F)
MDL: MFCD08669505
CAS-Nummer: 114012-41-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR