4-Keto-L-proline hydrobromide ≥90%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Oxo-L-proline hydrobromide

H57691.MD H57691.03
H57691.MDEA 45.8 EUR
H57691.MD H57691.03
4-Keto-L-proline hydrobromide ≥90%
4-Keto-L-proline hydrobromide

Purity: 90+% CAS No.: 75776-67-9 Molecular Formula: C5H7NO3.HBr Formula Weight: 210.03 Melting Point: 134-140° Optical Rotation: -32° (c=0.5 in methanol) MDL No.: MFCD12024572

Formel: C₅H₈BrNO₃
Molecular Weight: 210.03 g/mol
Schmelzpunkt: 134…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12024572
CAS-Nummer: 75776-67-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR