3-Oxetanol 95%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 3-Hydroxyoxetane

H57928.06 H57928.03
H57928.06EA 125 EUR
H57928.06 H57928.03
3-Oxetanol 95%
Oxetan-3-ol

Purity: 95% CAS No.: 7748-36-9 Molecular Formula: C3H6O2 Formula Weight: 74.08 Flash Point: 87°(189°F) Refractive Index: 1.438 Boiling Point: 153° MDL No.: MFCD09056790

Formel: C₃H₆O₂
Molecular Weight: 74.08 g/mol
Siedepunkt: 153 °C
Dichte: 1.167
Flammpunkt: 87 °C (189 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD09056790
CAS-Nummer: 7748-36-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR