2-n-Propyl-1-pentanol 97%
Lieferant: Thermo Scientific
|
Achtung
|
H57954.03
H57954.06
H57954.03EA
44.7
EUR
H57954.03
H57954.06
2-n-Propyl-1-pentanol 97%
2-n-Propyl-1-pentanol
Purity: 97%
CAS No.: 58175-57-8
Molecular Formula: C8H18O
Formula Weight: 130.23
Flash Point: 72°(162°F)
Physical Form: Liquid
Density: 0.83
Refractive Index: 1.4300
Appearance: Colourless to pale yellow
Boiling Point: 80-82°/12mm
MDL No.: MFCD00010260
Formel:
C₈H₁₈O Molecular Weight: 130,23 g/mol Siedepunkt: 80…82 °C (12 mmHg) Dichte: 0,83 Flammpunkt: 72 °C (162 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010260 CAS-Nummer: 58175-57-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...