3-Pyridin-4-ylpropionic acid 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-(4-Pyridyl)propionic Acid

H58655.03 H58655.06
H58655.03EA 148 EUR
H58655.03 H58655.06
3-Pyridin-4-ylpropionic acid 97%
3-Pyridin-4-ylpropionic acid

Purity: 97% CAS No.: 6318-43-0 Molecular Formula: C8H9NO2 Formula Weight: 151.16 Melting Point: 231° Beilstein Reference: 122234 MDL No.: MFCD00084846

Formel: C₈H₉NO₂
Molecular Weight: 151,17 g/mol
Schmelzpunkt: 231 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00084846
CAS-Nummer: 6318-43-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR